- Promotion of the transdermal delivery of protein drugs by
Promotion of the transdermal delivery of protein drugs by
International journal of nanomedicine (2016-11-09)
Ye Tu, Xinxia Wang, Ying Lu, He Zhang, Yuan Yu, Yan Chen, Junjie Liu, Zhiguo Sun, Lili Cui, Jing Gao, Yanqiang Zhong
PMID27822034
ABSTRACT
We recently reported that electret, which was prepared by a corona charging system with polypropylene film, could enhance the transdermal delivery of several drugs of low molecular weight. The aim of this study was to investigate whether electret could enhance the transdermal delivery of protein drugs by