MilliporeSigma
  • Total bile acids in the maternal and fetal compartment in relation to placental ABCG2 expression in preeclamptic pregnancies complicated by HELLP syndrome.
Total bile acids in the maternal and fetal compartment in relation to placental ABCG2 expression in preeclamptic pregnancies complicated by HELLP syndrome.
Biochimica et biophysica acta (2014-12-03)
Jiska Jebbink, Geertruda Veenboer, Souad Boussata, Remco Keijser, Andreas E Kremer, Ronald Oude Elferink, Joris van der Post, Gijs Afink, Carrie Ris-Stalpers
ABSTRACT
To investigate total bile acid (TBA) levels in maternal (MB) and umbilical cord blood (UCB) in normotensive, preeclamptic (PE), and PE pregnancies complicated by hemolysis elevated liver enzymes and low platelets (HELLP) syndrome in the context of ABCG2 placental gene expression levels, a recently reported placental bile acid transporter. TBA levels were determined in 83 paired MB and UCB samples of normotensive, PE and PE/HELLP pregnancies and in 22 paired arterial and venous UCB samples from uncomplicated term pregnancies. ABCG2 gene expression was measured in 104 human placentas by reverse transcriptase quantitative polymerase chain reaction. Overall, TBA levels in MB are higher compared to levels in UCB (p<0.0001), but this comparison looses statistical significance for the 11 PE/HELLP cases. TBA levels in maternal blood are increased in PE/HELLP compared to PE pregnancies (p=0.016). TBA levels in arterial and venous UCB from 22 normotensive pregnancies are not statistically different. ABCG2 expression is reduced in pregnancies where preeclampsia is further complicated by HELLP syndrome. ABCG2 expression in human placenta is not correlated with TBA levels in either the maternal or fetal compartment. Increased maternal TBA levels in PE/HELLP pregnancies indicate a relation between bile acids in the maternal circulation and HELLP syndrome. As overall TBA levels in maternal blood are increased compared to UCB, we conclude that the placenta partly protects the fetus from increased maternal TBA levels. This consistent difference in TBA levels between the maternal and fetal compartment is unrelated to the placental expression of ABCG2.
Sigma-Aldrich
HepaRG Cells BCRP (-/-), one vial
Pricing and availability is not currently available.