MilliporeSigma
  • Regulation of IL-20 Expression by Estradiol through KMT2B-Mediated Epigenetic Modification.
Regulation of IL-20 Expression by Estradiol through KMT2B-Mediated Epigenetic Modification.
PloS one (2016-11-03)
Chia-Hsin Su, I-Hsuan Lin, Tsai-Yu Tzeng, Wen-Ting Hsieh, Ming-Ta Hsu
ABSTRACT
Cytokines are low molecular weight regulatory proteins, or glycoproteins, with both tumor-promoting and inhibitory effects on breast cancer growth. Different cytokines play important roles in breast cancer initiation and progression. Here, we show that of the 39 interleukin (IL) genes, IL-20 is the only gene over-expressed in MCF-7 cells treated with estradiol (E2) and that induction of IL-20 expression by estrogen was epigenetically regulated. Methylation of histone H3K4 in the IL-20 promoter was shown to occur via the specific recruitment of KMT2B by estrogen receptor alpha (ERα), but not by other members of the mixed-lineage leukemia (MLL) family of histone methyltransferases. Depletion of KMT2B, or IL-20, disrupts estrogen signaling, attenuates cell proliferation, reduces colony formation, and results in cell cycle arrest. Furthermore, we demonstrated that KMT2B-mediated epigenetic modification also affected the expression of several ERα target genes. IL-20 and KMT2B expression were also associated with ERα-positive breast cancer tissues. We have revealed an important role for KMT2B in the epigenetic transcriptional regulation of cytokine IL-20, and other ERα-responsive genes, in breast cancer cells. Inhibition of IL-20 and KMT2B may have therapeutic benefits in ERα-positive breast cancer.
Sigma-Aldrich
Anti-monomethyl-Histone H3 (Lys4) Antibody, Upstate®, from rabbit
Pricing and availability is not currently available.
Sigma-Aldrich
MISSION® esiRNA, targeting human ESR1
Pricing and availability is not currently available.