MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • MicroRNA-455-3p modulates cartilage development and degeneration through modification of histone H3 acetylation.
MicroRNA-455-3p modulates cartilage development and degeneration through modification of histone H3 acetylation.
Biochimica et biophysica acta (2016-11-05)
Weishen Chen, Lingwu Chen, Ziji Zhang, Fangang Meng, Guangxin Huang, Puyi Sheng, Zhiqi Zhang, Weiming Liao
ABSTRACT
Histone acetylation regulated by class I histone deacetylases (HDACs) plays a pivotal role in matrix-specific gene transcription and cartilage development. While we previously demonstrated that microRNA (miR)-455-3p is upregulated during chondrogenesis and can enhance early chondrogenesis, the mechanism underlying this process remains largely unclear. In this study, we characterized the effect of miR-455-3p on histone H3 acetylation and its role during cartilage development and degeneration. We observed that miR-455-3p was highly expressed in proliferating and pre-hypertrophic chondrocytes, while HDAC2 and HDAC8 were primarily expressed in hypertrophic chondrocytes. Meanwhile, miR-455-3p suppressed the activity of reporter constructs containing the 3'-untranslated regions of HDAC2/8, inhibited HDAC2/8 expression and promoted histone H3 acetylation at the collagen 2 (COL2A1) promoter in human SW1353 chondrocyte-like cells. Treatment with the HDAC inhibitor trichostatin A (TSA) resulted in increased expression of cartilage-specific genes and promoted glycosaminoglycan deposition. Moreover, TSA inhibited matrix metalloproteinase 13 (Mmp13) expression and promoted nuclear translocation of SOX9 in interleukin-1-treated primary mouse chondrocytes. Lastly, knockdown of HDAC2/3/8 increased SRY (sex-determining region Y)-box 9 (SOX9) and decreased Runt-related transcription factor 2 (RUNX2) expression. Taken together, these findings suggest that miR-455-3p plays a critical role during chondrogenesis by directly targeting HDAC2/8 and promoting histone H3 acetylation, which raises possibilities of using miR-455-3p to influence chondrogenesis and cartilage degeneration.
Sigma-Aldrich
Magna ChIP® A/G Chromatin Immunoprecipitation Kit, Single day chromatin immunoprecipitation (ChIP) kit containing all necessary reagents to perform 22 individual chromatin immunoprecipitation (ChIP) reactions using magnetic A/G beads.