MilliporeSigma
  • An epitope-imprinted piezoelectric diagnostic tool for Neisseria meningitidis detection.
An epitope-imprinted piezoelectric diagnostic tool for Neisseria meningitidis detection.
Journal of molecular recognition : JMR (2016-08-05)
Neha Gupta, Kavita Shah, Meenakshi Singh
ABSTRACT
Neisseria meningitidis, a human-specific bacterial pathogen causes bacterial meningitis by invading the meninges (outer lining) of central nervous system. It is the polysaccharide present on the bacterial capsid that distinguishes various serogroups of N. meningitidis and can be utilized as antigens to elicit immune response. A computational approach identified candidate T-cell epitopes from outer membrane proteins Por B of N. meningitidis (MC58): (
Sigma-Aldrich
Ethylene glycol dimethacrylate, 98%, contains 90-110 ppm monomethyl ether hydroquinone as inhibitor