MilliporeSigma
  • Inhibition of hydrogen sulfide biosynthesis sensitizes lung adenocarcinoma to chemotherapeutic drugs by inhibiting mitochondrial DNA repair and suppressing cellular bioenergetics.
Inhibition of hydrogen sulfide biosynthesis sensitizes lung adenocarcinoma to chemotherapeutic drugs by inhibiting mitochondrial DNA repair and suppressing cellular bioenergetics.
Scientific reports (2016-11-04)
Bartosz Szczesny, Michela Marcatti, John R Zatarain, Nadiya Druzhyna, John E Wiktorowicz, Péter Nagy, Mark R Hellmich, Csaba Szabo
ABSTRACT
Therapeutic manipulation of the gasotransmitter hydrogen sulfide (H
Sigma-Aldrich
MISSION® esiRNA, targeting human CTH
Sigma-Aldrich
MISSION® esiRNA, targeting human CBS