MilliporeSigma
Production of xylonic acid by Klebsiella pneumoniae.
Applied microbiology and biotechnology (2016-09-16)
Chenhong Wang, Dong Wei, Zhongxi Zhang, Dexin Wang, Jiping Shi, Chul Ho Kim, Biao Jiang, Zengsheng Han, Jian Hao
ABSTRACT
The glucose oxidation pathway is important for glucose catabolism in Klebsiella pneumoniae. Gluconic acid and 2-ketogluconic acid are intermediates of this pathway, and the production of these two chemicals has been developed in K. pneumoniae mutants. Catalysis characteristic research in this study has shown that xylose is a suitable substrate of the glucose dehydrogenase of this pathway. Here, using xylose as substrate, xylonic acid was accumulated in the broth of K. pneumoniae culture, and this process was dependent upon acidic conditions. Using a mixture of glucose and xylose as substrates, a mixture of xylonic acid and gluconic acid was produced by the Δgad mutant of K. pneumoniae; gluconic acid was synthesized early, and xylonic acid synthesis began after most glucose was consumed. Using the hydrolysate of bamboo as substrate, mixture of 33 g/L gluconic acid and 14 g/L xylonic acid were produced by K. pneumoniae Δgad. In fed-batch fermentation, 103 g/L xylonic acid was produced after 79 h culture, with a conversion ratio of 1.11 g/g. This is the first report of xylonic acid produced by K. pneumoniae. Production of xylonic acid and gluconic acid using bamboo hydrolysate is a novel approach for biomass utilization.
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Sigma-Aldrich
L-(−)-Xylose, mixture of anomers, ≥99%