MilliporeSigma
  • Integrative genomics analysis identifies candidate drivers at 3q26-29 amplicon in squamous cell carcinoma of the lung.
Integrative genomics analysis identifies candidate drivers at 3q26-29 amplicon in squamous cell carcinoma of the lung.
Clinical cancer research : an official journal of the American Association for Cancer Research (2013-08-03)
Jing Wang, Jun Qian, Megan D Hoeksema, Yong Zou, Allan V Espinosa, S M Jamshedur Rahman, Bing Zhang, Pierre P Massion
ABSTRACT
Chromosome 3q26-29 is a critical region of genomic amplification in lung squamous cell carcinomas (SCC). Identification of candidate drivers in this region could help uncover new mechanisms in the pathogenesis and potentially new targets in SCC of the lung. We conducted a meta-analysis of seven independent datasets containing a total of 593 human primary SCC samples to identify consensus candidate drivers in 3q26-29 amplicon. Through integrating protein-protein interaction network information, we further filtered for candidates that may function together in a network. Computationally predicted candidates were validated using RNA interference (RNAi) knockdown and cell viability assays. Clinical relevance of the experimentally supported drivers was evaluated in an independent cohort of 52 lung SCC patients using survival analysis. The meta-analysis identified 20 consensus candidates, among which four (SENP2, DCUN1D1, DVL3, and UBXN7) are involved in a small protein-protein interaction network. Knocking down any of the four proteins led to cell growth inhibition of the 3q26-29-amplified SCC. Moreover, knocking down of SENP2 resulted in the most significant cell growth inhibition and downregulation of DCUN1D1 and DVL3. Importantly, a gene expression signature composed of SENP2, DCUN1D1, and DVL3 stratified patients into subgroups with different response to adjuvant chemotherapy. Together, our findings show that SENP2, DCUN1D1, and DVL3 are candidate driver genes in the 3q26-29 amplicon of SCC, providing novel insights into the molecular mechanisms of disease progression and may have significant implication in the management of SCC of the lung.
Sigma-Aldrich
Anti-SENP2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab2
Pricing and availability is not currently available.