MilliporeSigma
  • Simplified Enzyme-Linked Immunosorbent Assay for Routine Identification of Rhizobium japonicum Antigens.
Simplified Enzyme-Linked Immunosorbent Assay for Routine Identification of Rhizobium japonicum Antigens.
Applied and environmental microbiology (1985-04-01)
J Fuhrmann, A G Wollum
ABSTRACT
A simple, reliable, and flexible modification of the indirect enzyme-linked immunosorbent assay was developed for the identification of Rhizobium japonicum antigens from cultures and nodules. The procedure emphasizes efficient use of time and reagents, adaptability to variously equipped laboratories, and maintenance of sensitivity levels that are adequate for ecological studies.
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous glycerol solution