MilliporeSigma
  • AP-2 Is the Crucial Clathrin Adaptor Protein for CD4 Downmodulation by HIV-1 Nef in Infected Primary CD4+ T Cells.
AP-2 Is the Crucial Clathrin Adaptor Protein for CD4 Downmodulation by HIV-1 Nef in Infected Primary CD4+ T Cells.
Journal of virology (2015-10-02)
Marcos Vinicius Gondim, Linda Wiltzer-Bach, Brigitte Maurer, Carina Banning, Enrique Arganaraz, Michael Schindler
ABSTRACT
HIV-1 Nef-mediated CD4 downmodulation involves various host factors. We investigated the importance of AP-1, AP-2, AP-3, V1H-ATPase, β-COP, and ACOT8 for CD4 downmodulation in HIV-1-infected short hairpin RNA (shRNA)-expressing CD4(+) T cells and characterized direct interaction with Nef by Förster resonance energy transfer (FRET). Binding of lentiviral Nefs to CD4 and AP-2 was conserved, and only AP-2 knockdown impaired Nef-mediated CD4 downmodulation from primary T cells. Altogether, among the factors tested, AP-2 is the most important player for Nef-mediated CD4 downmodulation.