MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • E2/Estrogen receptor/sjogren syndrome-associated autoantigen relieves coactivator activator-induced G1/S arrest to promote breast tumorigenicity.
E2/Estrogen receptor/sjogren syndrome-associated autoantigen relieves coactivator activator-induced G1/S arrest to promote breast tumorigenicity.
Molecular and cellular biology (2014-02-26)
Yun Kyoung Kang, Sung Yun Jung, Jun Qin, Chao Li, Sophia Y Tsai, Ming-Jer Tsai, Bert W O'Malley
ABSTRACT
Coactivator activator (CoAA) is a dual-functional coregulator that regulates steroid receptor-mediated transcription and alternative splicing. Previously, we have shown that CoAA has tumor-suppressive potential in tumorigenic human kidney cells. Here, we uncover a molecular mechanism by which Sjogren syndrome-associated autoantigen (SSA), an estrogen receptor (ER) coactivator, induces MYC oncogene by removing repressive CoAA through E2-dependent degradation of CoAA and promotes G(1)/S transition of the cell cycle as well as anchorage-independent growth capability of breast cancer cells. We also show that E2 and ER enhance the E3 ligase activity of SSA to modulate CoAA through splicing isoform-selective ubiquitylation. We propose this as one potential molecular basis for the reduced tumor incidence in autoimmune disease patients and suggest SSA as a potential therapeutic target to treat breast cancer.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)