MilliporeSigma
  • A Drosophila homolog of bovine smg p25a GDP dissociation inhibitor undergoes a shift in isoelectric point in the developmental mutant quartet.
A Drosophila homolog of bovine smg p25a GDP dissociation inhibitor undergoes a shift in isoelectric point in the developmental mutant quartet.
Molecular and cellular biology (1993-01-01)
J E Zahner, C M Cheney
ABSTRACT
The Drosophila developmental mutation quartet causes late larval lethality and small imaginal discs and, when expressed in the adult female, has a lethal effect on early embryogenesis. These developmental defects are associated with mitotic defects, which include a low mitotic index in larval brains and incomplete separation of chromosomes in mitosis in the early embryo. quartet mutations also have a biochemical effect, i.e., a basic shift in isoelectric point in three proteins. We have purified one of these proteins, raised an antibody to it, and isolated and sequenced its cDNA. At the amino acid level, the sequence shows 68% identity and 81% similarity to bovine smg p25a GDP dissociation inhibitor (GDI), a regulator of ras-like small GTPases of the rab/SEC4/YPT1 subfamily. The correlation between a basic shift in isoelectric point in Drosophila GDI in quartet mutant tissue and the quartet developmental phenotype raises the possibility that a posttranslational modification of GDI is necessary for its function and that GDI function is essential for development.