MilliporeSigma
  • Adaptive regulation of human intestinal thiamine uptake by extracellular substrate level: a role for THTR-2 transcriptional regulation.
Adaptive regulation of human intestinal thiamine uptake by extracellular substrate level: a role for THTR-2 transcriptional regulation.
American journal of physiology. Gastrointestinal and liver physiology (2013-08-31)
Svetlana M Nabokina, Veedamali S Subramanian, Judith E Valle, Hamid M Said
ABSTRACT
The intestinal thiamine uptake process is adaptively regulated by the level of vitamin in the diet, but the molecular mechanism involved is not fully understood. Here we used the human intestinal epithelial Caco-2 cells exposed to different levels of extracellular thiamine to delineate the molecular mechanism involved. Our results showed that maintaining Caco-2 cells in a thiamine-deficient medium resulted in a specific and significant increase of [3H]thiamine uptake compared with cell exposure to a high level of thiamine (1 mM). This adaptive regulation was also associated with a higher level of mRNA expression of thiamine transporter-2 (THTR-2), but not thiamine transporter-1 (THTR-1), in the deficient condition and a higher level of promoter activity of gene encoding THTR-2 (SLC19A3). Using 5'-truncated promoter-luciferase constructs, we identified the thiamine level-responsive region in the SLC19A3 promoter to be between -77 and -29 (using transcriptional start site as +1). By means of mutational analysis, a key role for a stimulating protein-1 (SP1)/guanosine cytidine box in mediating the effect of extracellular thiamine level on SLC19A3 promoter was established. Furthermore, extracellular level of thiamine was found to affect SP1 protein expression and binding pattern to the thiamine level-responsive region of SLC19A3 promoter in Caco-2 cells as shown by Western blotting and electrophoretic mobility shift assay analysis, respectively. These studies demonstrate that the human intestinal thiamine uptake is adaptively regulated by the extracellular substrate level via transcriptional regulation of the THTR-2 system, and report that SP1 transcriptional factor is involved in this regulation.