MilliporeSigma
  • MicroRNA-146a Induced by Hypoxia Promotes Chondrocyte Autophagy through Bcl-2.
MicroRNA-146a Induced by Hypoxia Promotes Chondrocyte Autophagy through Bcl-2.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology (2015-10-23)
Fei Zhang, Jing Wang, Jianjun Chu, Cheng Yang, Hui Xiao, Chenglong Zhao, Zhengwang Sun, Xin Gao, Guanghui Chen, Zhitao Han, Weiwei Zou, Tielong Liu
ABSTRACT
There have been many studies on the etiology of osteoarthritis (OA) with regard to the function of inflammatory cytokines, the process of cartilage degradation, the function of miR-146a, hypoxia stimulation and autophagy in OA chondrocytes, but there have been no reports on the relationship between miR-146a and autophagy in cartilage, especially under hypoxia. This study aimed to confirm the relationship of miR-146a and autophagy in cartilage under hypoxia. Chondrocytes were treated by hypoxia gradients, and the main factors including HIF-1α, HIF-2α, miR-146a and Bcl-2 and autophagy markers ULK-1, ATG-5 were detected by quantitative PCR (Q-PCR) and western blotting. The autophagy marker LC-3 was detected by immunofluorescence. The reciprocal effects between miR-146a and Bcl-2 were confirmed by several combinations of shRNAs and adenovirus-gene systems followed by Q-PCR and western blot detection. Hypoxia maintained the chondrocytes phenotype and promoted autophagy and miR-146a expression via HIF-1α, but not HIF-2α, while miR-146a did not reversely affect HIF-1α. The autophagy induced by hypoxia through HIF-1α, miR-146a and Bcl-2. Simply, hypoxia induced HIF-1α, and HIF-1α increased miR-146a, but miR-146a suppressed Bcl-2, an autophagy inhibitor. While Bcl-2 affected neither HIF-1α nor miR-146a. The absence of both HIF-1α and miR-146a or Bcl-2 over-expression inhibited hypoxia-induced autophagy. HIF-1α, miR-146a and Bcl-2 play crucial roles during hypoxia-induced autophagy, Hypoxia, HIF-1α and miR-146a promote chondrocytes autophagy via depressing Bcl-2. We conclude that miR-146a may serve as a novel therapeutic target for protecting cartilage from degeneration in OA.