MilliporeSigma
  • Immunolocalization of CB1 receptor in rat striatal neurons: a confocal microscopy study.
Immunolocalization of CB1 receptor in rat striatal neurons: a confocal microscopy study.
Synapse (New York, N.Y.) (2004-07-06)
F R Fusco, A Martorana, C Giampà, Z De March, D Farini, V D'Angelo, G Sancesario, G Bernardi
ABSTRACT
Several lines of evidence indicate that cannabinoids, among other functions, are involved in motor control. Although cannabinoid receptors (CB(1)) mRNA has been observed in medium-sized spiny neurons of the striatum, a description of the precise localization of CB(1) at a protein level among striatal cells is still lacking. Therefore, we performed immunohistochemical studies with light and confocal microscopy to identify neuronal subpopulations that express CB(1) and to assess the distribution of the receptor within these neurons. In our single label light microscopy study, CB(1) was observed in most medium-sized neurons of the caudate-putamen. However, CB(1) was also present in large-sized neurons scattered throughout the striatum. Our dual-label study showed that 89.3% of projection neurons in matrix contain CB(1), and that 56.4% of projection neurons in patch are labeled for CB(1). To investigate the presence of CB(1) among the different subclasses of striatal interneurons we performed a double-labeling study matching CB(1) and each of the striatal interneuron markers, namely, choline acetyl-transferase, parvalbumin, calretinin, and nitric oxide synthase. Our double-label study showed that most parvalbumin immunoreactive interneurons (86.5%), more than one-third (39.2%) of cholinergic interneurons, and about one-third (30.4%) of the NOS-positive neurons are labeled for CB(1). Calretinin-immunolabeled neurons were devoid of CB(1).