MilliporeSigma
A plasma membrane Ca2+ ATPase isoform at the postsynaptic density.
Neuroscience (2010-08-04)
A C Burette, E E Strehler, R J Weinberg
ABSTRACT
Most excitatory input in the hippocampus impinges on dendritic spines. Entry of Ca(2+) into spines through NMDA receptors can trigger a sequence of biochemical reactions leading to sustained changes in synaptic efficacy. To provide specificity, dendritic spines restrict the diffusion of Ca(2+) signaling and downstream molecules. The postsynaptic density (PSD) (the most prominent subdomain within the spine) is the site of Ca(2+) entry through NMDA receptors. We here demonstrate that Ca(2+) can also be removed via pumps embedded in the PSD. Using light- and electron-microscopic immunohistochemistry, we find that PMCA2w, a member of the plasma membrane Ca(2+)-ATPase (PMCA) family, concentrates at the PSD of most hippocampal spines. We propose that PMCA2w may be recruited into supramolecular complexes at the postsynaptic density, thus helping to regulate Ca(2+) nanodomains at subsynaptic sites. Taken together, these results suggest a novel function for PMCAs as modulators of Ca(2+) signaling at the synapse.
Sigma-Aldrich
Anti-Vesicular Glutamate Transporter 1 Antibody, serum, Chemicon®