MilliporeSigma
  • Developmental competence of prepubertal goat oocytes selected with brilliant cresyl blue and matured with cysteamine supplementation.
Developmental competence of prepubertal goat oocytes selected with brilliant cresyl blue and matured with cysteamine supplementation.
Reproduction, Nutrition, Development (2003)
Rodriguez-Gonzalez E, et al.