MilliporeSigma
  • Vitrification of immature and matured bovine oocytes: effect of brilliant cresyl blue selection and hyaluronan addition.
Vitrification of immature and matured bovine oocytes: effect of brilliant cresyl blue selection and hyaluronan addition.
Animal Reproduction (2016)