MilliporeSigma
  • Degradation of myoglobin by polymeric artificial metalloproteases containing catalytic modules with various catalytic group densities: site selectivity in peptide bond cleavage.
Degradation of myoglobin by polymeric artificial metalloproteases containing catalytic modules with various catalytic group densities: site selectivity in peptide bond cleavage.
Journal of the American Chemical Society (2003-11-20)
Chang Eun Yoo, Pil Seok Chae, Jung Eun Kim, Eui June Jeong, Junghun Suh
ABSTRACT
Mononuclear, dinuclear, and tetranuclear artificial metalloproteases were prepared by attaching respective catalytic modules containing the Cu(II) complex of cyclen (Cu(II)Cyc) to a derivative of cross-linked polystyrene. The polymeric artificial metalloproteases effectively cleaved peptide bonds of myoglobin (Mb) by hydrolysis. The proteolytic activity increased considerably as the catalytic group density was raised: the ratio of k(cat)/K(m) was 1:13:100 for the mono-, di-, and tetranuclear catalysts. In the degradation of Mb by the dinuclear catalyst, two pairs of intermediate proteins accumulated. One of the two initial cleavage sites leading to the formation of the protein fragments is identified as Gln(91)-Ser(92) and the other is suggested as Ala(94)-Thr(95). On the basis of a molecular modeling study by using the X-ray crystallographic structure of Mb, the site-selectivity is attributed to anchorage of one Cu(II)Cyc unit of the catalytic module to a heme carboxylate of Mb. The high site selectivity for the initial cleavage of a protein substrate and mechanistic analysis of the catalytic action are unprecedented for polymeric artificial enzymes.