MilliporeSigma
  • Spectrophotometric determination of gold(III) in forensic and pharmaceutical samples and results complemented with ICP AES and EDXRF analysis.
Spectrophotometric determination of gold(III) in forensic and pharmaceutical samples and results complemented with ICP AES and EDXRF analysis.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2016-10-05)
Vani Nagaraja, M Kiran Kumar, Nagendrappa Giddappa
ABSTRACT
Spectrophotometric method with three systems were developed here for the determination of gold(III) using o-dianisidine, aniline sulphate and catechol. Gold(III),in the system 1 it oxidizes o-dianisidine, in the system 2 it oxidizes catechol followed by its coupling with o-dianisidine, in the system 3 it oxidizes catechol followed by its coupling with aniline sulphate forming dye products with respective λmax 446nm, 540nm, and 505nm. All the three systems were optimized and analytical parameters were calculated. The molar absorptivity values were 9.27×104, 1.97×104 and 1.62×104 respectively for the systems 1, 2 and 3 with the corresponding Sandell sensitivity values (μgcm-2), 0.0021, 0.0096 and 0.011. The optimized systems were used for the determination of gold present in some forensic jewellery and pharmaceutical samples and the results obtained were compared with the results of all samples determined by Inductively Coupled Plasma - Atomic Emission Spectrometric method and a few of them were also complemented by Energy Dispersive X-Ray Fluorescent spectral analysis.