MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Diosmin-induced senescence, apoptosis and autophagy in breast cancer cells of different p53 status and ERK activity.
Diosmin-induced senescence, apoptosis and autophagy in breast cancer cells of different p53 status and ERK activity.
Toxicology letters (2016-11-29)
Anna Lewinska, Jagoda Adamczyk-Grochala, Ewa Kwasniewicz, Anna Deregowska, Maciej Wnuk
ABSTRACT
Relatively low bioavailability of plant-derived nutraceuticals with anticancer properties may limit their usefulness for prevention and therapy of cancer. In the present study, we have screened for nutraceuticals (n=30) that would act at low micromolar range against phenotypically distinct breast cancer cell lines, namely MCF-7 (ER+, PR+/-, HER2-), MDA-MB-231 (ER-, PR-, HER2-) and SK-BR-3 (ER-, PR-, HER2+), and diosmin, a citrus fruit flavonoid belonging to a flavone subclass, was selected. MCF-7 cell line was found to be the most sensitive to diosmin treatment. Diosmin caused G2/M cell cycle arrest, elevation in p53, p21 and p27 levels and stress-induced premature senescence when used at lower concentrations (5 and 10μM). Diosmin (20μM) also promoted apoptosis that was not observed in normal human mammary epithelial cells (HMEC). Diosmin stimulated oxidative and nitrosative stress, DNA damage and changes in global DNA methylation patterns. The status of p53 (wild type versus mutant) and the levels of phosphorylated ERK1/2 in a steady state, and diosmin-induced autophagy may reflect diverse response to diosmin treatment in MCF-7, MDA-MB-231 and SK-BR-3 cells, which in turn results in different cell fates. Taken together, diosmin that acts at low micromolar range against breast cancer cells may be considered as a promising candidate for anticancer therapy.
Sigma-Aldrich
MAHMA NONOate