MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • PI3K Pathway Inhibition Achieves Potent Antitumor Activity in Melanoma Brain Metastases In Vitro and In Vivo.
PI3K Pathway Inhibition Achieves Potent Antitumor Activity in Melanoma Brain Metastases In Vitro and In Vivo.
Clinical cancer research : an official journal of the American Association for Cancer Research (2016-11-03)
Heike Niessner, Jennifer Schmitz, Ghazaleh Tabatabai, Andreas M Schmid, Carsten Calaminus, Tobias Sinnberg, Benjamin Weide, Thomas K Eigentler, Claus Garbe, Birgit Schittek, Leticia Quintanilla-Fend, Benjamin Bender, Marion Mai, Christian Praetorius, Stefan Beissert, Gabriele Schackert, Michael H Muders, Matthias Meinhardt, Gustavo B Baretton, Reinhard Dummer, Keith Flaherty, Bernd J Pichler, Dagmar Kulms, Dana Westphal, Friedegund Meier
ABSTRACT
Great advances have recently been made in treating patients with metastatic melanoma. However, existing therapies are less effective on cerebral than extracerebral metastases. This highlights the potential role of the brain environment on tumor progression and drug resistance and underlines the need for "brain-specific" therapies. We previously showed that the PI3K-AKT survival pathway is hyperactivated in brain but not extracerebral melanoma metastases and that astrocyte-conditioned medium activates AKT in melanoma cells in vitro We therefore tested the PI3K inhibitor buparlisib as an antitumor agent for melanoma brain metastases. Buparlisib inhibited AKT activity, decreased proliferation, and induced apoptosis in metastatic melanoma cell lines and short-term brain melanoma cells, irrespective of their BRAF and NRAS mutation status. In addition, buparlisib inhibited hyperactivated AKT and induced apoptosis in melanoma cells that were stimulated with astrocyte-conditioned medium. The growth of tumors induced by injecting human BRAF- and NRAS-mutant metastatic melanoma cells into the brain of mice was significantly inhibited by buparlisib. These results emphasize the value of targeting the PI3K pathway as a strategy to develop drugs for melanoma brain metastases. Clin Cancer Res; 22(23); 5818-28. ©2016 AACR.
Sigma-Aldrich
Suplatast tosylate, ≥98% (HPLC)