MilliporeSigma
  • Identification of candidate domestication regions in the radish genome based on high-depth resequencing analysis of 17 genotypes.
Identification of candidate domestication regions in the radish genome based on high-depth resequencing analysis of 17 genotypes.
TAG. Theoretical and applied genetics. Theoretische und angewandte Genetik (2016-07-06)
Namshin Kim, Young-Min Jeong, Seongmun Jeong, Goon-Bo Kim, Seunghoon Baek, Young-Eun Kwon, Ara Cho, Sang-Bong Choi, Jiwoong Kim, Won-Jun Lim, Kyoung Hyoun Kim, Won Park, Jae-Yoon Kim, Jin-Hyun Kim, Bomi Yim, Young Joon Lee, Byung-Moon Chun, Young-Pyo Lee, Beom-Seok Park, Hee-Ju Yu, Jeong-Hwan Mun
ABSTRACT
This study provides high-quality variation data of diverse radish genotypes. Genome-wide SNP comparison along with RNA-seq analysis identified candidate genes related to domestication that have potential as trait-related markers for genetics and breeding of radish. Radish (Raphanus sativus L.) is an annual root vegetable crop that also encompasses diverse wild species. Radish has a long history of domestication, but the origins and selective sweep of cultivated radishes remain controversial. Here, we present comprehensive whole-genome resequencing analysis of radish to explore genomic variation between the radish genotypes and to identify genetic bottlenecks due to domestication in Asian cultivars. High-depth resequencing and multi-sample genotyping analysis of ten cultivated and seven wild accessions obtained 4.0 million high-quality homozygous single-nucleotide polymorphisms (SNPs)/insertions or deletions. Variation analysis revealed that Asian cultivated radish types are closely related to wild Asian accessions, but are distinct from European/American cultivated radishes, supporting the notion that Asian cultivars were domesticated from wild Asian genotypes. SNP comparison between Asian genotypes identified 153 candidate domestication regions (CDRs) containing 512 genes. Network analysis of the genes in CDRs functioning in plant signaling pathways and biochemical processes identified group of genes related to root architecture, cell wall, sugar metabolism, and glucosinolate biosynthesis. Expression profiling of the genes during root development suggested that domestication-related selective advantages included a main taproot with few branched lateral roots, reduced cell wall rigidity and favorable taste. Overall, this study provides evolutionary insights into domestication-related genetic selection in radish as well as identification of gene candidates with the potential to act as trait-related markers for background selection of elite lines in molecular breeding.