MilliporeSigma
  • Strained silacycles in organic synthesis: a new reagent for the enantioselective allylation of aldehydes.
Strained silacycles in organic synthesis: a new reagent for the enantioselective allylation of aldehydes.
Journal of the American Chemical Society (2002-07-04)
James W A Kinnaird, Pui Yee Ng, Katsumi Kubota, Xiaolun Wang, James L Leighton
ABSTRACT
A new reagent for the enantioselective allylation of aliphatic aldehydes has been developed. The reagent is easily prepared in a single step from commercially available materials and may be stored without significant decomposition. The reactivity of the reagent is attributed to strain-release Lewis acidity.