MilliporeSigma
  • Synthesis, characterization, and application of novel trihexyl tetradecyl phosphonium bis(2,4,4-trimethylpentyl)phosphinate for extractive desulfurization of liquid fuel.
Synthesis, characterization, and application of novel trihexyl tetradecyl phosphonium bis(2,4,4-trimethylpentyl)phosphinate for extractive desulfurization of liquid fuel.
Fuel Processing Technology (2014)
Dharaskar SA, et al.