MilliporeSigma
  • Pyrrolidine dithiocarbamate inhibits NF-kappa B mobilization and TNF production in human monocytes.
Pyrrolidine dithiocarbamate inhibits NF-kappa B mobilization and TNF production in human monocytes.
Journal of Immunology (1993)
Ziegler-Heitbrock HW, et al.