MilliporeSigma
  • Human and murine class I MHC antigens share conserved serine 335, the site of HLA phosphorylation in vivo.
Human and murine class I MHC antigens share conserved serine 335, the site of HLA phosphorylation in vivo.
The Journal of Biological Chemistry (1984)
Guild BC and Strominger JL