MilliporeSigma
  • Functional consequences of preorganized helical structure in the intrinsically disordered cell-cycle inhibitor p27(Kip1).
Functional consequences of preorganized helical structure in the intrinsically disordered cell-cycle inhibitor p27(Kip1).
Biochemistry (2002)
Bienkiewicz EA, et al.