MilliporeSigma
  • Silver stain for proteins in polyacrylamide gels: a modified procedure with enhanced uniform sensitivity.
Silver stain for proteins in polyacrylamide gels: a modified procedure with enhanced uniform sensitivity.
Analytical Biochemistry (1981)
Morrissey JH.