MilliporeSigma
  • Coordinated Activities of Multiple Myc-dependent and Myc-independent Biosynthetic Pathways in Hepatoblastoma.
Coordinated Activities of Multiple Myc-dependent and Myc-independent Biosynthetic Pathways in Hepatoblastoma.
The Journal of biological chemistry (2016-10-16)
Huabo Wang, Jie Lu, Lia R Edmunds, Sucheta Kulkarni, James Dolezal, Junyan Tao, Sarangarajan Ranganathan, Laura Jackson, Marc Fromherz, Donna Beer-Stolz, Radha Uppala, Sivakama Bharathi, Satdarshan P Monga, Eric S Goetzman, Edward V Prochownik
ABSTRACT
Hepatoblastoma (HB) is associated with aberrant activation of the β-catenin and Hippo/YAP signaling pathways. Overexpression of mutant β-catenin and YAP in mice induces HBs that express high levels of c-Myc (Myc). In light of recent observations that Myc is unnecessary for long-term hepatocyte proliferation, we have now examined its role in HB pathogenesis using the above model. Although Myc was found to be dispensable for in vivo HB initiation, it was necessary to sustain rapid tumor growth. Gene expression profiling identified key molecular differences between myc