MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Fixed Versus Variable Dosing of Prothrombin Complex Concentrate in Vitamin K Antagonist-Related Intracranial Hemorrhage: A Retrospective Analysis.
Fixed Versus Variable Dosing of Prothrombin Complex Concentrate in Vitamin K Antagonist-Related Intracranial Hemorrhage: A Retrospective Analysis.
Neurocritical care (2016-04-08)
Rahat Amadkhan Abdoellakhan, Ishita Parveen Miah, Nakisa Khorsand, Karina Meijer, Korné Jellema
ABSTRACT
Millions of patients receive vitamin K antagonist (VKA) therapy worldwide. Annually 0.2-1 % of all VKA users develops an intracranial hemorrhage (ICH). Prothrombin complex concentrate (PCC) is administered to restore the INR ≤ 1.5 in an attempt to limit hematoma growth. In order to facilitate PCC dosing, our hospital recently changed from a variable dose based on bodyweight, baseline- and target-INR, to a fixed 1000 IU fIX PCC dosing protocol for ICH. In a before and after design, we compared successful achievement of an INR ≤ 1.5 with a fixed dosing strategy versus the variable dosing strategy of PCC in patients presenting with intracranial bleeding complications of VKA. Data of the two cohorts of patients were retrospectively collected from medical records. A median dosage of 1750 IU was given per patient in the variable dose group (n = 25) versus 1000 IU in the fixed dose group (n = 28). In the intention-to-treat analysis, 96 % achieved an INR ≤ 1.5 after an initial dose in the variable dose cohort compared to 68 % in the fixed dose cohort (p = 0.01). An additional dose was given in 2 (8 %) versus 9 (32 %) patients, respectively (p = 0.04). The median door-to-PCC-order time was 42 versus 32 min (p = 0.37) and the door-to-needle time was 81, respectively 60 min (p = 0.42). The fixed dose protocol necessitates additional PCC infusions more frequently to achieve a target INR ≤ 1.5. Door-to-order and door-to-needle time were shorter but, in this small cohort, not significantly so. The effect on clinical outcome remains unknown.
Sigma-Aldrich
Pyridinium chlorochromate, 98%