MilliporeSigma
  • A practical synthesis of ethyl (R)-and (S)-2-hydroxy-4-phenylbutanoate and D-homophenylalanine ethyl ester hydrochloride from L-malic acid.
A practical synthesis of ethyl (R)-and (S)-2-hydroxy-4-phenylbutanoate and D-homophenylalanine ethyl ester hydrochloride from L-malic acid.
Tetrahedron Asymmetry (2001)
Lin WQ, et al.