MilliporeSigma
  • Ethylene and carbon dioxide production by developing strawberries show a correlative pattern that is indicative of ripening climacteric fruit.
Ethylene and carbon dioxide production by developing strawberries show a correlative pattern that is indicative of ripening climacteric fruit.
Physiologia Plantarum (2006)
Lannetta PM et al