MilliporeSigma
  • Use of tRNA consensus primers to indicate subgroups of Pseudomonas solanacearum by polymerase chain reaction amplification.
Use of tRNA consensus primers to indicate subgroups of Pseudomonas solanacearum by polymerase chain reaction amplification.
Applied and Environmental Microbiology (1992)
Seal SE et al