MilliporeSigma
  • Evaluation of respiration with clark type electrode in isolated mitochondria and permeabilized animal cells.
Evaluation of respiration with clark type electrode in isolated mitochondria and permeabilized animal cells.
Methods in Molecular Biology (2012)
Silva AM and Oliveira PJ