MilliporeSigma
  • Low oxygen tension during in vitro maturation is beneficial for supporting the subsequent development of bovine cumulus-oocyte complexes.
Low oxygen tension during in vitro maturation is beneficial for supporting the subsequent development of bovine cumulus-oocyte complexes.
Molecular Reproduction and Development (2000)
Hashimoto S et al.