MilliporeSigma
  • Genetic diversity within Streptococcus mutans evident from chromosomal DNA restriction fragment polymorphisms.
Genetic diversity within Streptococcus mutans evident from chromosomal DNA restriction fragment polymorphisms.
Journal of Clinical Microbiology (1989)
Caufield PW and Walker TM