MilliporeSigma
  • A rapid procedure for isolating chromosomal DNA from Lactobacillus species and other Gram-positive bacteria.
A rapid procedure for isolating chromosomal DNA from Lactobacillus species and other Gram-positive bacteria.
Letters in Applied Microbiology (2001)
Ulrich RL and Hughes TA