MilliporeSigma
  • Angiotensin II induces interleukin-1?-mediated islet inflammation and ?-cell dysfunction independently of vasoconstrictive effects.
Angiotensin II induces interleukin-1?-mediated islet inflammation and ?-cell dysfunction independently of vasoconstrictive effects.
Diabetes (2015)
Sauter NS et al.