MilliporeSigma
  • Biomarkers of cigarette smoking and DNA methylating agents: Raman, SERS and DFT study of 3-methyladenine and 7-methyladenine.
Biomarkers of cigarette smoking and DNA methylating agents: Raman, SERS and DFT study of 3-methyladenine and 7-methyladenine.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2017-01-07)
Scott G Harroun, Yaoting Zhang, Tzu-Heng Chen, Ching-Rong Ku, Huan-Tsung Chang
ABSTRACT
3-Methyladenine and 7-methyladenine are biomarkers of DNA damage from exposure to methylating agents. For example, the concentration of 3-methyladenine increases significantly in the urine of cigarette smokers. Surface-enhanced Raman spectroscopy (SERS) has shown much potential for detection of biomolecules, including DNA. Much work has been dedicated to the canonical nucleobases, with comparatively fewer investigations of modified DNA and modified DNA nucleobases. Herein, Raman spectroscopy and SERS are used to examine the adsorption orientations of 3-methyladenine and 7-methyladenine on Ag nanoparticles. Density functional theory (DFT) calculations at the B3LYP level are used to support the conclusions via simulated spectra of the nucleobases and of Ag
Sigma-Aldrich
7-Methyladenine, 97%
Sigma-Aldrich
3-Methyladenine, autophagy inhibitor