MilliporeSigma
  • Acetylation of aromatic cysteine conjugates by recombinant human N-acetyltransferase 8.
Acetylation of aromatic cysteine conjugates by recombinant human N-acetyltransferase 8.
Xenobiotica; the fate of foreign compounds in biological systems (2016-05-18)
Reema Deol, P David Josephy
ABSTRACT
1. The mercapturic acid (MA) pathway is a metabolic route for the processing of glutathione conjugates to MA (N-acetylcysteine conjugates). An N-acetyltransferase enzyme, NAT8, catalyzes the transfer of an acetyl group from acetyl-CoA to the cysteine amino group, producing a MA, which is excreted in the urine. We expressed human NAT8 in HEK293T cells and developed an HPLC-MS method for the quantitation of the S-aryl-substituted cysteine conjugates and their MA. 2. We measured the activity of the enzyme for acetylation of benzyl-, 4-nitrobenzyl-, and 1-menaphthylcysteine substrates. 3. NAT8 catalyzed the acetylation of all three cysteine conjugates with similar Michaelis-Menten kinetics.
Sigma-Aldrich
S-Benzyl-L-cysteine, 97%