MilliporeSigma
  • Mammalian WDR12 is a novel member of the Pes1-Bop1 complex and is required for ribosome biogenesis and cell proliferation.
Mammalian WDR12 is a novel member of the Pes1-Bop1 complex and is required for ribosome biogenesis and cell proliferation.
The Journal of cell biology (2005-07-27)
Michael Hölzel, Michaela Rohrmoser, Martin Schlee, Thomas Grimm, Thomas Harasim, Anastassia Malamoussi, Anita Gruber-Eber, Elisabeth Kremmer, Wolfgang Hiddemann, Georg W Bornkamm, Dirk Eick
ABSTRACT
Target genes of the protooncogene c-myc are implicated in cell cycle and growth control, yet the linkage of both is still unexplored. Here, we show that the products of the nucleolar target genes Pes1 and Bop1 form a stable complex with a novel member, WDR12 (PeBoW complex). Endogenous WDR12, a WD40 repeat protein, is crucial for processing of the 32S precursor ribosomal RNA (rRNA) and cell proliferation. Further, a conditionally expressed dominant-negative mutant of WDR12 also blocks rRNA processing and induces a reversible cell cycle arrest. Mutant WDR12 triggers accumulation of p53 in a p19ARF-independent manner in proliferating cells but not in quiescent cells. Interestingly, a potential homologous complex of Pes1-Bop1-WDR12 in yeast (Nop7p-Erb1p-Ytm1p) is involved in the control of ribosome biogenesis and S phase entry. In conclusion, the integrity of the PeBoW complex is required for ribosome biogenesis and cell proliferation in mammalian cells.
Sigma-Aldrich
Monoclonal Anti-BOP1 antibody produced in rat, ~1.0 mg/mL, clone BOP 6H11, purified immunoglobulin, buffered aqueous solution