MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Improving the Performance of Formamidinium and Cesium Lead Triiodide Perovskite Solar Cells using Lead Thiocyanate Additives.
Improving the Performance of Formamidinium and Cesium Lead Triiodide Perovskite Solar Cells using Lead Thiocyanate Additives.
ChemSusChem (2016-10-27)
Yue Yu, Changlei Wang, Corey R Grice, Niraj Shrestha, Jing Chen, Dewei Zhao, Weiqiang Liao, Alexander J Cimaroli, Paul J Roland, Randy J Ellingson, Yanfa Yan
ABSTRACT
Formamidinium lead triiodide (FAPbI
Sigma-Aldrich
4-tert-Butylpyridine, 98%