MilliporeSigma
  • Upregulation of miR-146a by YY1 depletion correlates with delayed progression of prostate cancer.
Upregulation of miR-146a by YY1 depletion correlates with delayed progression of prostate cancer.
International journal of oncology (2017-01-20)
Yeqing Huang, Tao Tao, Chunhui Liu, Han Guan, Guangyuan Zhang, Zhixin Ling, Lei Zhang, Kai Lu, Shuqiu Chen, Bin Xu, Ming Chen
ABSTRACT
Previously published studies explained that the excessive expression of miR-146a influences the prostate cancer (PCa) cells in terms of apoptosis, progression, and viability. Although miR-146a acts as a tumor suppressor, current knowledge on the molecular mechanisms that controls its expression in PCa is limited. In this study, gene set enrichment analysis (GSEA) showed negatively enriched expression of miR-146a target gene sets and positively enriched expression of gene sets suppressed by the enhancer of zeste homolog 2 (EZH2) after YY1 depletion in PCa cells. The current results demonstrated that the miR-146a levels in PCa tissues with high Gleason scores (>7) are significantly lower than those in PCa tissues with low Gleason scores (≤7), which were initially observed in the clinical specimens. An inverse relationship between YY1 and miR-146a expression was also observed. Experiments indicated the decrease in cell viability, proliferation, and promoting apoptosis after YY1 depletion, while through inhibiting miR-146a could alleviate the negative effect brought by YY1 depletion. We detected the reversed adjustment of YY1 to accommodate miR-146a transcriptions. On the basis of YY1 depletion, we determined that the expression of miR-146a increased after EZH2 knockdown. We validated the combination of YY1 and its interaction with EZH2 at the miR-146a promoter binding site, thereby prohibiting the transcriptional activity of miR-146a in PCa cells. Our results suggested that YY1 depletion repressed PCa cell viability and proliferation and induced apoptosis at least in a miR-146a-assisted manner.
Sigma-Aldrich
DL-Glyceraldehyde, ≥90% (GC)
Sigma-Aldrich
MISSION® esiRNA, targeting human YY1