MilliporeSigma
  • Ultra-thin layer chromatography with integrated silver colloid-based SERS detection.
Ultra-thin layer chromatography with integrated silver colloid-based SERS detection.
Electrophoresis (2016-08-16)
Ryan A Wallace, Nickolay V Lavrik, Michael J Sepaniak
ABSTRACT
Simplified lab-on-a-chip techniques are desirable for quick and efficient detection of analytes of interest in the field. The following work involves the use of deterministic pillar arrays on the micro-scale as a platform to separate compounds, and the use of Ag colloid within the arrays as a source of increased signal via surface enhanced Raman spectroscopy (SERS). One problem traditionally seen with SERS surfaces containing Ag colloid is oxidation; however, our platforms are superhydrophobic, reducing the amount of oxidation taking place on the surface of the Ag colloid. This work includes the successful separation and SERS detection of a fluorescent dye compounds (resorufin and sulforhodamine 640), fluorescent anti-tumor drugs (Adriamycin and Daunomycin), and purine and pyrimidine bases (adenine, cytosine, guanine, hypoxanthine, and thymine).
Sigma-Aldrich
Trichloro(octadecyl)silane, ≥90%