MilliporeSigma
  • The drug candidate, ADX71441, is a novel, potent and selective positive allosteric modulator of the GABA
The drug candidate, ADX71441, is a novel, potent and selective positive allosteric modulator of the GABA
Neuropharmacology (2016-11-28)
Mikhail Kalinichev, Françoise Girard, Hasnaà Haddouk, Mélanie Rouillier, Eric Riguet, Isabelle Royer-Urios, Vincent Mutel, Robert Lütjens, Sonia Poli
ABSTRACT
Positive allosteric modulation of the GABA