MilliporeSigma
  • α-Defensin 5 gene expression is regulated by gut microbial metabolites.
α-Defensin 5 gene expression is regulated by gut microbial metabolites.
Bioscience, biotechnology, and biochemistry (2016-10-21)
Yutaka Sugi, Kyoko Takahashi, Kenta Kurihara, Kou Nakano, Tetsuro Kobayakawa, Kazuaki Nakata, Masato Tsuda, Shigemasa Hanazawa, Akira Hosono, Shuichi Kaminogawa
ABSTRACT
α-Defensin 5 is important to both maintenance of a gut microbiota and host immunity. While previous reports have shown that gut bacteria are able to upregulate α-defensin 5 through Toll-like receptor signaling, we demonstrate here that α-defensin 5 expression can also be regulated by microbial metabolites. Among these, lactate appeared to significantly suppress α-defensin 5 gene transcription. Actually, fractions of <3 kD compounds obtained from the ceca of SPF mice were suppressed α-defensin 5 gene transcription at specific concentrations. Our results also suggest that cecal content may include as yet unidentified factors that can enhance α-defensin 5 expression. Our data point to a novel function for the gut microbial metabolites in controlling the expression of antimicrobial peptides in the intestine.