- Disruption of quercetin metabolism by fungicide affects energy production in honey bees (
Disruption of quercetin metabolism by fungicide affects energy production in honey bees (
Proceedings of the National Academy of Sciences of the United States of America (2017-02-15)
Wenfu Mao, Mary A Schuler, May R Berenbaum
PMID28193870
ABSTRACT
Cytochrome P450 monooxygenases (P450) in the honey bee,