MilliporeSigma
  • Evaluation of impact of social support and care on HIV-positive and AIDS individuals' quality of life: a nonrandomised community trial.
Evaluation of impact of social support and care on HIV-positive and AIDS individuals' quality of life: a nonrandomised community trial.
Journal of clinical nursing (2016-09-21)
Xing-Ming Li, Xiao-Qing Yuan, Jun-Jie Wang, Wan-Ying Zhang, Yang Zhou, Gu-Ning Liu
ABSTRACT
Our study was conducted to further investigate the model of social support and care for People Living with HIV/AIDS(PLHA), to explore their role in People Living with AIDS's quality of life (QOL) as reference for improving nursing policies for AIDS. Social support and care are the most important factors impacting the QOL of People Living with HIV/AIDS, but most studies conducted upon the influence of social support and QOL of People Living with HIV/AIDS are mainly based on cross-sectional design. Our study was a nonrandomised controlled community intervention study. The participants diagnosed as People Living with HIV/AIDS at Beijing You An Hospital received a comprehensive social support care from December 2013 to December 2014. To evaluate the impact of social support and care model on People Living with HIV/AIDS, our study analysed the different dimension scores of social support scale and quality of life before and after the intervention. Correlation between the net benefit value of social support and that of QOL from various dimensions were analysed. There were significant differences in the score of objective support and usage of support (all p = 0·02) for social support. Net values of objective support score and usage of support were 0·25 and 0·19, respectively, after intervention. There were significant differences in physiological function, role physical, general health, vitality, social function, mental health, health transition and total score of quality of life (all p < 0·05). The canonical correlation analysis of net values of social support and QOL indicated that the first and second canonical correlation were statistically significant, with correlation coefficients of 0·53 (p = 0·00) and 0·21 (p = 0·04). Social support and care intervention model can effectively improve perceived subjective feeling on social support and QOL condition for People Living with HIV/AIDS. And strategies to improve social support and care intervention programmes are strongly encouraged. The method is simple and cost-effective and could be a way to improve the quality of life condition for People Living with HIV/AIDS.