MilliporeSigma
  • Novel glutamate receptor antagonists selectively protect against kainic acid neurotoxicity in cultured cerebral cortex neurons.
Novel glutamate receptor antagonists selectively protect against kainic acid neurotoxicity in cultured cerebral cortex neurons.
Journal of neurochemistry (1990-11-01)
A Frandsen, P Krogsgaard-Larsen, A Schousboe
ABSTRACT
The effect on excitatory amino acid (EAA)-induced toxicity of two novel non-N-methyl-D-aspartate (non-NMDA) antagonists 2-amino-3-[3-(carboxymethoxy)-5-methylisoxazol-4-yl]propionic acid (AMOA) and 2-amino-3-[2-(3-hydroxy-5-methyl-isoxazol-4-yl)methyl-5-methyl-3- oxoisoxazolin-4-yl]propionic acid (AMNH) was tested in primary cultures of cerebral cortex neurons. Such cultures provide a useful model for the investigation of the toxicity of EAAs and a convenient screening system for potential neuroprotective activity of pharmacological agents. It was demonstrated that AMNH and AMOA abolished neurotoxicity induced by kainic acid with IC50 values of 62 +/- 10 and 120 +/- 19 microM, respectively. No effect on neuronal damage induced by NMDA or AMPA could be detected.